Learn More
Uline Extra Long Tabletop Impulse Sealer with Cutter - 24 | | 1 EA
Catalog No. NC2753786
Encompass
Seal and trim larger bags and poly tubing in one step. Gently press to seal then slide cutter across tubing to create a perfect sized bag. Recommended for jumbo items - up to 24 wide. Delivers consistent quality for high-volume applications. Rugged and durable construction. No warmup time needed. 8-step adjustable timer. Replacement Blades and Service Kit with replacement heating element available.
Catalog No. NC2753786 Supplier Uline Supplier No. H1254
Seal and trim larger bags and poly tubing in one step. Gently press to seal then slide cutter across tubing to create a perfect sized bag. Recommended for jumbo items - up to 24 wide. Delivers consistent quality for high-volume applications. Rugged and durable construction. No warmup time needed. 8-step adjustable timer. Replacement Blades and Service Kit with replacement heating element available.