Learn More
Apexbio Technology LLC XL019 945755-56-6 10mM (in 1mL DMSO)
SDP
Catalog No. 5000064348
Encompass_Preferred
XL019 (CAS 945755-56-6) is a selective inhibitor targeting Janus kinase 2 (JAK2) with an IC50 of 2 2 nM JAK2 regulates signaling pathways controlling erythroid granulocytic and megakaryocytic lineage differentiation via cytokine and hematopoietic growth factor receptors including erythropoietin and thrombopoietin receptors XL019 demonstrates high selectivity for JAK2 over related kinases (e g JAK1 TYK2) In mouse models bearing HEL 92 1 7 xenografts oral administration of XL019 significantly reduced tumor growth XL019 is evaluated clinically as a candidate for treating primary myelofibrosis and secondary myelofibrosis associated with polycythemia vera or essential thrombocythemia
Catalog No. 50-000-64348 Supplier Apexbio Technology LLC Supplier No. A39375.1
XL019 (CAS 945755-56-6) is a selective inhibitor targeting Janus kinase 2 (JAK2) with an IC50 of 2 2 nM JAK2 regulates signaling pathways controlling erythroid granulocytic and megakaryocytic lineage differentiation via cytokine and hematopoietic growth factor receptors including erythropoietin and thrombopoietin receptors XL019 demonstrates high selectivity for JAK2 over related kinases (e g JAK1 TYK2) In mouse models bearing HEL 92 1 7 xenografts oral administration of XL019 significantly reduced tumor growth XL019 is evaluated clinically as a candidate for treating primary myelofibrosis and secondary myelofibrosis associated with polycythemia vera or essential thrombocythemia