Learn More
Medchemexpress LLC HY-101973 1mg Medchemexpress, XL041 CAS:1256918-39-4 Purity:>98%
SDP
Catalog No. 501967941
Encompass_Preferred
Medchemexpress, HY-101973 1mg XL041 CAS:1256918-39-4 XL041 (BMS-852927) is an LXRβ-selective agonist. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-7941 Supplier Medchemexpress LLC Supplier No. HY1019731MG
Medchemexpress, HY-101973 1mg XL041 CAS:1256918-39-4 XL041 (BMS-852927) is an LXRβ-selective agonist. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.