Learn More
Agilent Technologies XL1-BLUE ELECTROCOMP CELLS
Catalog No. 50125045
Encompass
XL1-Blue Electroporation-Competent Cells are optimal hosts for propagating both lambda and plasmid vectors. They are endA minus cells, Improve the quality of miniprep DNA, allow for color screening and single-strand rescue of phagemid DNA
Catalog No. 50-125-045 Supplier Agilent Technologies Supplier No. 200228
XL1-Blue Electroporation-Competent Cells are optimal hosts for propagating both lambda and plasmid vectors. They are endA minus cells, Improve the quality of miniprep DNA, allow for color screening and single-strand rescue of phagemid DNA