Learn More
Medchemexpress LLC XL177A | 2417089-74-6 | 99.6% | 861.47 | 50 MG
SDP
Catalog No. 5000349271
Encompass_Preferred
XL177A is a highly potent and selective irreversible USP7 inhibitor with an IC50 of 0.34 nM, demonstrating cancer cell killing through a p53-dependent mechanism. It acts as a potent USP7 inhibitor and p53 stabilizer in vitro, effectively suppressing cancer cell growth. The compound selectively labels the catalytic cysteine, C223, of USP7 across the DUBome and human proteome.
  • Highly potent and selective irreversible USP7 inhibitor (IC50 of 0.34 nM).
  • Elicits cancer cell killing via a p53-dependent mechanism.
  • Suppresses cancer cell growth predominantly through p53-dependent pathways.
  • Labels USP7's catalytic cysteine (C223) with exquisite selectivity.
  • Induces complete G1 arrest in MCF7 cells after 24 hours (at 1 μM).
  • Promotes rapid degradation of HDM2, increasing p53 and p21 protein levels in MCF7 cells.
Catalog No. 50-003-49271 Supplier Medchemexpress LLC Supplier No. HY13879450MG
XL177A is a highly potent and selective irreversible USP7 inhibitor with an IC50 of 0.34 nM, demonstrating cancer cell killing through a p53-dependent mechanism. It acts as a potent USP7 inhibitor and p53 stabilizer in vitro, effectively suppressing cancer cell growth. The compound selectively labels the catalytic cysteine, C223, of USP7 across the DUBome and human proteome.
  • Highly potent and selective irreversible USP7 inhibitor (IC50 of 0.34 nM).
  • Elicits cancer cell killing via a p53-dependent mechanism.
  • Suppresses cancer cell growth predominantly through p53-dependent pathways.
  • Labels USP7's catalytic cysteine (C223) with exquisite selectivity.
  • Induces complete G1 arrest in MCF7 cells after 24 hours (at 1 μM).
  • Promotes rapid degradation of HDM2, increasing p53 and p21 protein levels in MCF7 cells.