Learn More
Selleck Chemical LLC XL177A S8967-10MM/1ML
SDP
Catalog No. 5000025089
Encompass_Preferred
XL177A is a potent selective and irreversible inhibitor of Ubiquitin specific peptidase 7 (USP7) with IC50 of 0 34 nM
Catalog No. 50-000-25089 Supplier Selleck Chemical LLC Supplier No. S896710MM/1ML
XL177A is a potent selective and irreversible inhibitor of Ubiquitin specific peptidase 7 (USP7) with IC50 of 0 34 nM