Learn More
Selleck Chemical LLC XL177A-5mg
SDP
Supplier:  Selleck Chemical LLC S89675MG
Encompass_Preferred
XL177A is a potent, selective and irreversible inhibitor of Ubiquitin specific peptidase 7 (USP7) with IC50 of 0.34 nM.
Catalog No. 50-238-9744