Learn More
Medchemexpress LLC HY-138794 5mg , XL177A CAS:2417089-74-6 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1387945MG
Encompass_Preferred
HY-138794 5mg XL177A CAS: 2417089-74-6 XL177A is a highly potent and selective irreversible USP7 inhibitor with an IC50 of 0.34 nM. XL177A elicits cancer cell killing through a p53-dependent mechanism. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-225-9356