Learn More
Medchemexpress LLC Xl177a USP7 inhibitor | 2417089-74-6 | 99.6% | C48H57ClN8O5 | 10MG
SDP
Catalog No. 5000260353
Encompass_Preferred
A potent, selective, irreversible inhibitor of ubiquitin-specific peptidase 7 (USP7), characterized by sub-nanomolar biochemical potency and cellular activity that stabilizes p53 and suppresses cancer cell growth through a p53-dependent mechanism. Supplied as a research reagent with high reported purity and available in solid and solution formats for biochemical and cellular studies.
  • Sub-nanomolar biochemical potency (IC50 0.34 nM) against USP7.
  • Selective, irreversible mechanism of inhibition.
  • Elicits cancer cell killing predominantly via a p53-dependent pathway.
  • High purity suitable for research applications (reported 99.6%).
  • Available as solid and DMSO solution formats for assay flexibility.
  • Useful as a biochemical probe for USP7 function and p53-related cancer research.
Catalog No. 50-002-60353 Supplier Medchemexpress LLC Supplier No. HY13879410MG
A potent, selective, irreversible inhibitor of ubiquitin-specific peptidase 7 (USP7), characterized by sub-nanomolar biochemical potency and cellular activity that stabilizes p53 and suppresses cancer cell growth through a p53-dependent mechanism. Supplied as a research reagent with high reported purity and available in solid and solution formats for biochemical and cellular studies.
  • Sub-nanomolar biochemical potency (IC50 0.34 nM) against USP7.
  • Selective, irreversible mechanism of inhibition.
  • Elicits cancer cell killing predominantly via a p53-dependent pathway.
  • High purity suitable for research applications (reported 99.6%).
  • Available as solid and DMSO solution formats for assay flexibility.
  • Useful as a biochemical probe for USP7 function and p53-related cancer research.