Learn More
Agilent Technologies XL2-BLUE MRF ULTRACOMP CELLS
Catalog No. 50125017
Encompass
XL2-Blue MRF' Ultracompetent Cells deficient in all known E. coli K12 restriction systems. Increase the efficiency of introducing eukaryotic DNA into E. coli, mcrA, mcrCB and mrr mutations, prevent cleavage of methylated DNA.
Catalog No. 50-125-017 Supplier Agilent Technologies Supplier No. 200151
XL2-Blue MRF' Ultracompetent Cells deficient in all known E. coli K12 restriction systems. Increase the efficiency of introducing eukaryotic DNA into E. coli, mcrA, mcrCB and mrr mutations, prevent cleavage of methylated DNA.