Learn More
Agilent Technologies XL2-BLUE ULTRACOMPETENT CELLS
Supplier:  Agilent Technologies 200150
Encompass
Cell, Ultracompetent, XL2-Blue; Stratagene; Highest cloning efficiency, greater than equal5 x 109 transformants/ug; Preparation of quality plasmid DNA; Blue-white color screening; Contain antibiotic-resistant F episome; Versatile cloning
Catalog No. 50-125-016