Learn More
Apexbio Technology LLC XL335 629664-81-9 10mM (in 1mL DMSO)
SDP
Catalog No. 5000063790
Encompass_Preferred
XL335 (CAS 629664-81-9) is a selective orally bioavailable agonist of the farnesoid X receptor (FXR) displaying an EC50 of approximately 4 nM In Hep3B liver cancer cells XL335 activation of FXR reduces interleukin-6-induced C-reactive protein (CRP) expression at both mRNA and protein levels In vivo studies indicate that XL335 diminishes lipopolysaccharide (LPS)-stimulated inflammatory responses and fructose-induced hepatic inflammation by suppressing acute-phase proteins (e g SAP SAA3) and adipose differentiation-related protein (ADRP) Thus XL335 serves as a valuable tool for investigating FXR-mediated pathways involved in liver inflammation and metabolic disorders
Catalog No. 50-000-63790 Supplier Apexbio Technology LLC Supplier No. B15285.1
XL335 (CAS 629664-81-9) is a selective orally bioavailable agonist of the farnesoid X receptor (FXR) displaying an EC50 of approximately 4 nM In Hep3B liver cancer cells XL335 activation of FXR reduces interleukin-6-induced C-reactive protein (CRP) expression at both mRNA and protein levels In vivo studies indicate that XL335 diminishes lipopolysaccharide (LPS)-stimulated inflammatory responses and fructose-induced hepatic inflammation by suppressing acute-phase proteins (e g SAP SAA3) and adipose differentiation-related protein (ADRP) Thus XL335 serves as a valuable tool for investigating FXR-mediated pathways involved in liver inflammation and metabolic disorders