Learn More
Medchemexpress LLC HY-13806 5mg Medchemexpress, XL388 CAS:1251156-08-7 Purity:>98%
SDP
Catalog No. 502081516
Encompass_Preferred
Medchemexpress, HY-13806 5mg XL388 CAS:1251156-08-7 XL388 is a highly potent and ATP-competitive mTOR inhibitor with an IC50 of 9.9 nM. XL388 simultaneously inhibits both mTORC1 and mTORC2. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-1516 Supplier Medchemexpress LLC Supplier No. HY138065MG
Medchemexpress, HY-13806 5mg XL388 CAS:1251156-08-7 XL388 is a highly potent and ATP-competitive mTOR inhibitor with an IC50 of 9.9 nM. XL388 simultaneously inhibits both mTORC1 and mTORC2. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.