Learn More
Medchemexpress LLC HY-15260A 100mg , XL413 (hydrochloride) CAS:1169562-71-3 Purity:98%
SDP
Catalog No. 501493178
Encompass_Preferred
Medchemexpress, HY-15260A 100mg XL413 (hydrochloride) CAS:1169562-71-3 Formula:C14H13Cl2N3O2 IC50: 3.4 nM (Cdc7), 42 nM (PIM1), 215 nM (CK2) EC50: 118 nM (pMCM) Purity:98% XL413 hydrochloride is a potent, selective and ATP competitive inhibitor of Cdc7 , with an IC 50 of 3.4 nM, and also shows potent effect with IC 50 s of 215, 42 nM on CK2, PIM1, respectively, and an EC 50 of 118 nM on pMCM. Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-149-3178 Supplier Medchemexpress LLC Supplier No. HY15260A100MG
Medchemexpress, HY-15260A 100mg XL413 (hydrochloride) CAS:1169562-71-3 Formula:C14H13Cl2N3O2 IC50: 3.4 nM (Cdc7), 42 nM (PIM1), 215 nM (CK2) EC50: 118 nM (pMCM) Purity:98% XL413 hydrochloride is a potent, selective and ATP competitive inhibitor of Cdc7 , with an IC 50 of 3.4 nM, and also shows potent effect with IC 50 s of 215, 42 nM on CK2, PIM1, respectively, and an EC 50 of 118 nM on pMCM. Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.