Learn More
Medchemexpress LLC XL44 | 2637949-91-6 | 98.0% | 388.39 | 25 MG
SDP
Catalog No. 5000394301
Encompass_Preferred
XL44, an hRpn13 binder, induces hRpn13-dependent apoptosis and also restricts cell viability by a PCLAF-dependent mechanism. It induces ubiquitin-dependent loss of hRpn13Pru and ubiquitin-independent loss of select KEN box containing proteins.
  • Induces hRpn13-dependent apoptosis
  • Restricts cell viability by a PCLAF-dependent mechanism
  • Induces ubiquitin-dependent loss of hRpn13Pru
  • Induces ubiquitin-independent loss of select KEN box containing proteins
Catalog No. 50-003-94301 Supplier Medchemexpress LLC Supplier No. 25MGHY162393
XL44, an hRpn13 binder, induces hRpn13-dependent apoptosis and also restricts cell viability by a PCLAF-dependent mechanism. It induces ubiquitin-dependent loss of hRpn13Pru and ubiquitin-independent loss of select KEN box containing proteins.
  • Induces hRpn13-dependent apoptosis
  • Restricts cell viability by a PCLAF-dependent mechanism
  • Induces ubiquitin-dependent loss of hRpn13Pru
  • Induces ubiquitin-independent loss of select KEN box containing proteins