Learn More
Medchemexpress LLC Xl888 | 1149705-71-4 | MFCD22124888 | 99.4% | C29H37N5O3 | 10MG
SDP
Catalog No. 5000261676
Encompass_Preferred
XL888 is an orally bioavailable, ATP-competitive small-molecule inhibitor of heat shock protein 90 (HSP90) used in oncology research. It destabilizes HSP90 client proteins, leading to cell cycle arrest and apoptosis, and has demonstrated anti-proliferative and anti-tumor activity in preclinical studies. Reported biochemical potency against HSP90 is ~24 nM.
  • Orally bioavailable small-molecule suitable for in vivo and in vitro studies.
  • ATP-competitive mechanism that targets HSP90 chaperone activity.
  • Demonstrated anti-proliferative and pro-apoptotic effects in cancer cell lines.
  • Reported low nanomolar biochemical potency in cell-free assays.
  • Useful for pharmacology and mechanism-of-action studies involving HSP90 clients.
Catalog No. 50-002-61676 Supplier Medchemexpress LLC Supplier No. HY1331310MG
XL888 is an orally bioavailable, ATP-competitive small-molecule inhibitor of heat shock protein 90 (HSP90) used in oncology research. It destabilizes HSP90 client proteins, leading to cell cycle arrest and apoptosis, and has demonstrated anti-proliferative and anti-tumor activity in preclinical studies. Reported biochemical potency against HSP90 is ~24 nM.
  • Orally bioavailable small-molecule suitable for in vivo and in vitro studies.
  • ATP-competitive mechanism that targets HSP90 chaperone activity.
  • Demonstrated anti-proliferative and pro-apoptotic effects in cancer cell lines.
  • Reported low nanomolar biochemical potency in cell-free assays.
  • Useful for pharmacology and mechanism-of-action studies involving HSP90 clients.