Learn More
Medchemexpress LLC HY-13313 5mg Medchemexpress, XL888 CAS:1149705-71-4 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY133135MG
Encompass_Preferred
Medchemexpress, HY-13313 5mg XL888 CAS:1149705-71-4 XL888 is a heat shock protein-90 (HSP90) inhibitor, with an IC50 of 24 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-1331