Learn More
Electron Microscopy Sciences X-Large Bulk Specimen Holder
SDP
Catalog No. 501927140
Encompass_Preferred
Holds up to 2" specimens. Opening: 52 mm (2") wide x 40 mm (1 1/2") long x 13 mm ( 1/2") deep. Pin Diameter: 3.2 mm ( 1/8"). Material: Machined aluminum with stainless steel allen set screws. Allen wrench included.
This item is not returnable. View return policy
Catalog No. 50-192-7140 Supplier Electron Microscopy Sciences Supplier No. 7595108
Holds up to 2" specimens. Opening: 52 mm (2") wide x 40 mm (1 1/2") long x 13 mm ( 1/2") deep. Pin Diameter: 3.2 mm ( 1/8"). Material: Machined aluminum with stainless steel allen set screws. Allen wrench included.