Learn More
NEOGEN Xylose Lysine Deoxycholate (XLD) Agar, 5 kg
SDP
Catalog No. 502015232
Encompass_Preferred
Xylose Lysine Deoxycholate Agar is used for the isolation and differentiation of enteric pathogens. Conforms to Harmonized USP/EP/JP Requirements. XLD Agar is not intended for use in the diagnosis of disease or other conditions in humans. A medium recommended by the Harmonized Pharmacopeia for isolation and identification of Salmonella from non-sterile products. Conforms to USP/EP/JP performance specification. Originally formulated by Taylor to differentiate enteric pathogens, the agar is widely used as the preferred differential medium for Salmonella spp. The medium is void of peptones but instead uses yeast extract as a carbon, nitrogen and vitamin source and xylose, lactose and sucrose are fermentable carbohydrates.For more information visit https://www.neogen.com/solutions/Microbiology/xylose-lysine-deoxycholate-xld-agar
Catalog No. 50-201-5232 Supplier NEOGEN Supplier No. 700003041
Xylose Lysine Deoxycholate Agar is used for the isolation and differentiation of enteric pathogens. Conforms to Harmonized USP/EP/JP Requirements. XLD Agar is not intended for use in the diagnosis of disease or other conditions in humans. A medium recommended by the Harmonized Pharmacopeia for isolation and identification of Salmonella from non-sterile products. Conforms to USP/EP/JP performance specification. Originally formulated by Taylor to differentiate enteric pathogens, the agar is widely used as the preferred differential medium for Salmonella spp. The medium is void of peptones but instead uses yeast extract as a carbon, nitrogen and vitamin source and xylose, lactose and sucrose are fermentable carbohydrates.For more information visit https://www.neogen.com/solutions/Microbiology/xylose-lysine-deoxycholate-xld-agar