Learn More
NEOGEN Xylose Lysine Deoxycholate (XLD) Agar (ISO), 500 g
SDP
Catalog No. 502015213
Encompass_Preferred
This item has a minimum order quantity of 2 per supplier requirements.
Xylose Lysine Deoxycholate (XLD) Agar (ISO) is a selective agar for the detection of Salmonella spp. in food, animal feed and in environmental samples from the food production area as described in ISO 6579-1:2017. Xylose Lysine Deoxycholate (XLD) Agar (ISO) is not intended for use in the diagnosis of disease or other conditions in humans. A selective, diagnostic agar for the detection of Salmonella spp. in food, animal feed and in environmental samples from the food production area as described in ISO 6579-1:2017. Originally formulated by Taylor to differentiate enteric pathogens, the agar is widely used as the preferred differential medium for Salmonella spp .For more information visit https://www.neogen.com/solutions/Microbiology/xylose-lysine-deoxycholate-xld-agar-iso
Catalog No. 50-201-5213 Supplier NEOGEN Supplier No. 700004393
Add to Cart
Add to Cart
This item has a minimum order quantity of 2 per supplier requirements.
Xylose Lysine Deoxycholate (XLD) Agar (ISO) is a selective agar for the detection of Salmonella spp. in food, animal feed and in environmental samples from the food production area as described in ISO 6579-1:2017. Xylose Lysine Deoxycholate (XLD) Agar (ISO) is not intended for use in the diagnosis of disease or other conditions in humans. A selective, diagnostic agar for the detection of Salmonella spp. in food, animal feed and in environmental samples from the food production area as described in ISO 6579-1:2017. Originally formulated by Taylor to differentiate enteric pathogens, the agar is widely used as the preferred differential medium for Salmonella spp .For more information visit https://www.neogen.com/solutions/Microbiology/xylose-lysine-deoxycholate-xld-agar-iso