Learn More
NEOGEN Xylose Lysine Deoxycholate (XLD) Agar (ISO), 5 kg
SDP
Catalog No. 502015214
Encompass_Preferred
Xylose Lysine Deoxycholate (XLD) Agar (ISO) is a selective agar for the detection of Salmonella spp. in food, animal feed and in environmental samples from the food production area as described in ISO 6579-1:2017. Xylose Lysine Deoxycholate (XLD) Agar (ISO) is not intended for use in the diagnosis of disease or other conditions in humans. A selective, diagnostic agar for the detection of Salmonella spp. in food, animal feed and in environmental samples from the food production area as described in ISO 6579-1:2017. Originally formulated by Taylor to differentiate enteric pathogens, the agar is widely used as the preferred differential medium for Salmonella spp. The medium is void of peptones but instead uses yeast extract as a carbon, nitrogen and vitamin source and xylose, lactose and sucrose are fermentable carbohydrates.For more information visit https://www.neogen.com/solutions/Microbiology/xylose-lysine-deoxycholate-xld-agar-iso
Catalog No. 50-201-5214 Supplier NEOGEN Supplier No. 700004394
Xylose Lysine Deoxycholate (XLD) Agar (ISO) is a selective agar for the detection of Salmonella spp. in food, animal feed and in environmental samples from the food production area as described in ISO 6579-1:2017. Xylose Lysine Deoxycholate (XLD) Agar (ISO) is not intended for use in the diagnosis of disease or other conditions in humans. A selective, diagnostic agar for the detection of Salmonella spp. in food, animal feed and in environmental samples from the food production area as described in ISO 6579-1:2017. Originally formulated by Taylor to differentiate enteric pathogens, the agar is widely used as the preferred differential medium for Salmonella spp. The medium is void of peptones but instead uses yeast extract as a carbon, nitrogen and vitamin source and xylose, lactose and sucrose are fermentable carbohydrates.For more information visit https://www.neogen.com/solutions/Microbiology/xylose-lysine-deoxycholate-xld-agar-iso