Learn More
MOLTOX Molecular Toxicology Inc XLD Agar, 20 mL/plate, 10 plates/sleeve, Store in Refrigerator
SDP
Catalog No. 50948816
Encompass_Preferred
Xylose Lysine Desoxycholate Agar (XLD) is a selective medium for the isolation and differentiation of enteric pathogens, particularly the more fastidious pathogens such as Shigella. Each lot of media from MOLTOX is tested for sterility, where applicable, and its ability to support the growth of the appropriate microorganism. All media are accompanied by GLP-compliant formulation and Quality Control certificate.
Catalog No. 50-948-816 Supplier MOLTOX Molecular Toxicology Inc Supplier No. 5140S016
Xylose Lysine Desoxycholate Agar (XLD) is a selective medium for the isolation and differentiation of enteric pathogens, particularly the more fastidious pathogens such as Shigella. Each lot of media from MOLTOX is tested for sterility, where applicable, and its ability to support the growth of the appropriate microorganism. All media are accompanied by GLP-compliant formulation and Quality Control certificate.