Learn More
MSC Excel Dryer XLERATOR Hand Dryer, Chrome Plated, Automatic, Adjustable Heat
Catalog No. 502838628
Encompass
The XLERATOR hand dryer is a high-speed, automatic unit designed for efficient and hygienic hand drying. The XL-C model features a durable chrome-plated cover and provides adjustable speed, sound, and heat control, making it suitable for various commercial environments while offering significant cost and energy savings.
  • 8 second dry time.
  • Adjustable speed and sound control.
  • Adjustable heat control (high, medium, low, off).
  • Automatic sensor operated activation.
  • Chrome plated cover for durability.
  • Achieves over 95% cost savings compared to paper towels.
  • Uses up to 80% less energy than conventional hand dryers.
  • Equipped with a HEPA filtration system that removes 99.999% of viruses from the airstream.
  • Includes a 7-year limited warranty.
  • Designed for surface mounting.
Catalog No. 50-283-8628 Supplier MSC Supplier No. 38866083
The XLERATOR hand dryer is a high-speed, automatic unit designed for efficient and hygienic hand drying. The XL-C model features a durable chrome-plated cover and provides adjustable speed, sound, and heat control, making it suitable for various commercial environments while offering significant cost and energy savings.
  • 8 second dry time.
  • Adjustable speed and sound control.
  • Adjustable heat control (high, medium, low, off).
  • Automatic sensor operated activation.
  • Chrome plated cover for durability.
  • Achieves over 95% cost savings compared to paper towels.
  • Uses up to 80% less energy than conventional hand dryers.
  • Equipped with a HEPA filtration system that removes 99.999% of viruses from the airstream.
  • Includes a 7-year limited warranty.
  • Designed for surface mounting.