Learn More
MSC Excel Dryer Hand Dryer, Chrome Plated, HEPA Filter, 110-120V
Catalog No. 502838215
Encompass
This XLERATOR Hand Dryer is engineered for high-speed, hygienic hand drying. It provides an 8-second dry time and incorporates a HEPA filtration system that removes 99.999% of viruses from the airstream. Designed for energy efficiency, it offers substantial savings compared to conventional hand dryers and paper towels.
  • Adjustable speed and sound control
  • Adjustable heat settings
  • HEPA filter installed
  • 8 second dry time
  • Less than 1% failure rate
  • 80% less energy than conventional hand dryers
  • 95% cost savings versus paper towels
  • Helps qualify for several LEED credits
  • Noise reduction nozzle
  • 7-year limited warranty
Catalog No. 50-283-8215 Supplier MSC Supplier No. 38866075
This XLERATOR Hand Dryer is engineered for high-speed, hygienic hand drying. It provides an 8-second dry time and incorporates a HEPA filtration system that removes 99.999% of viruses from the airstream. Designed for energy efficiency, it offers substantial savings compared to conventional hand dryers and paper towels.
  • Adjustable speed and sound control
  • Adjustable heat settings
  • HEPA filter installed
  • 8 second dry time
  • Less than 1% failure rate
  • 80% less energy than conventional hand dryers
  • 95% cost savings versus paper towels
  • Helps qualify for several LEED credits
  • Noise reduction nozzle
  • 7-year limited warranty