Learn More
MSC Excel Dryer Hand Dryer, White Thermoset Resin, 8 Second Dry Time, Automatic Sensor Operated
Catalog No. 502837437
Encompass
The XLERATOR Hand Dryer offers a high-performance, energy-efficient, and hygienic solution for commercial and public restrooms. It provides a fast dry time and significant cost savings, contributing to environmental sustainability by reducing paper towel waste.
  • Dries hands in 8 seconds
  • Features adjustable speed, sound, and heat control
  • Delivers up to 95% cost savings compared to paper towels
  • Contributes to LEED credits and reduces carbon footprint
  • Available with a HEPA filtration system to remove 99.999% of viruses from the airstream
  • Automatic sensor-operated and surface-mounted for easy installation
  • Includes a 7-year limited warranty
Catalog No. 50-283-7437 Supplier MSC Supplier No. 38866539
The XLERATOR Hand Dryer offers a high-performance, energy-efficient, and hygienic solution for commercial and public restrooms. It provides a fast dry time and significant cost savings, contributing to environmental sustainability by reducing paper towel waste.
  • Dries hands in 8 seconds
  • Features adjustable speed, sound, and heat control
  • Delivers up to 95% cost savings compared to paper towels
  • Contributes to LEED credits and reduces carbon footprint
  • Available with a HEPA filtration system to remove 99.999% of viruses from the airstream
  • Automatic sensor-operated and surface-mounted for easy installation
  • Includes a 7-year limited warranty