Learn More
MSC Excel Dryer Hand Dryer, 1450 Watt, 110-120V, White Epoxy
Supplier: MSC 38865952
The XLERATOR Hand Dryer is a high-speed, energy-efficient unit for various applications. Operating at 110-120V, it features a 1450 Watt heating element and a white epoxy finish. This automatic, sensor-operated dryer includes an eHEPA filtration system, offering hygienic drying with adjustable speed and heat controls.
- 8 second dry time.
- Adjustable speed and sound control.
- Adjustable heat control (high, medium, low, and off).
- Externally visible service LED.
- Less than 1% failure rate.
- eHEPA filtration removes 99.999% of viruses and 99.97% of bacteria.
- 80% less energy than conventional hand dryers.
- 95% cost savings versus paper towels.
- Building Green approved and helps qualify for LEED credits.
- Antimicrobial wall guards designed to protect walls.
- Automatic sensor operated.
- Surface mounted.
- ADA-compliant with recess kit.