Learn More
MSC Excel Dryer Electric Hand Dryer, 1490 Watt, White Finish, 208/277 V
Catalog No. 502837910
Encompass
This electric hand dryer features a 1490 Watt motor and a clean white finish, designed for efficient and rapid hand drying. It operates on 208/277 V power and includes a noise reduction nozzle for quieter operation in various environments. The compact design ensures easy installation in a range of washroom settings.
  • Powerful 1490 Watt operation
  • Compatible with 208/277 V electrical systems
  • Features a noise reduction nozzle
  • Measures approximately 11-3/4 in wide x 12-11/16 in high x 6-11/16 in deep for compact installation.
Catalog No. 50-283-7910 Supplier MSC Supplier No. 38866687
This electric hand dryer features a 1490 Watt motor and a clean white finish, designed for efficient and rapid hand drying. It operates on 208/277 V power and includes a noise reduction nozzle for quieter operation in various environments. The compact design ensures easy installation in a range of washroom settings.
  • Powerful 1490 Watt operation
  • Compatible with 208/277 V electrical systems
  • Features a noise reduction nozzle
  • Measures approximately 11-3/4 in wide x 12-11/16 in high x 6-11/16 in deep for compact installation.