Learn More
MSC Excel Dryer XLERATOR Hand Dryer, 1490 W, 208/277 V, Custom Image Finish
Catalog No. 502838631
Encompass
The XLERATOR Hand Dryer is an automatic sensor-operated, high-speed unit designed for efficient and hygienic hand drying. It offers significant cost and energy savings compared to paper towels, contributing to a reduced carbon footprint. This model features a custom image finish and includes a HEPA filter, ensuring advanced air hygiene.
  • 8 second dry time
  • Adjustable speed and sound control
  • Adjustable heat control: high, medium, low, and off
  • Externally visible service LED
  • 95% cost savings versus paper towels
  • 80% less energy than conventional hand dryers
  • HEPA filtration system removes 99.999% of virus from the airstream
  • Contributes to WELL points and LEED credits
  • 7-year limited warranty
Catalog No. 50-283-8631 Supplier MSC Supplier No. 38866463
The XLERATOR Hand Dryer is an automatic sensor-operated, high-speed unit designed for efficient and hygienic hand drying. It offers significant cost and energy savings compared to paper towels, contributing to a reduced carbon footprint. This model features a custom image finish and includes a HEPA filter, ensuring advanced air hygiene.
  • 8 second dry time
  • Adjustable speed and sound control
  • Adjustable heat control: high, medium, low, and off
  • Externally visible service LED
  • 95% cost savings versus paper towels
  • 80% less energy than conventional hand dryers
  • HEPA filtration system removes 99.999% of virus from the airstream
  • Contributes to WELL points and LEED credits
  • 7-year limited warranty