Learn More
MSC Excel Dryer Hand Dryer, Custom Special Image Cover, 208-277V, 1490W
Catalog No. 502838212
Encompass
The XLERATOR hand dryer offers an 8-second dry time and custom image cover for personalized branding. Operating at 208-277V and 1490W, it provides energy and cost savings over paper towels, supporting environmental goals.
  • 8-second dry time
  • Adjustable speed, sound, and heat control
  • Visible service LED
  • 95% cost savings vs. paper towels
  • Reduces carbon footprint up to 88% vs. paper towels
  • Building Green Approved; contributes to WELL points & LEED credits
  • Optional electrostatic HEPA filtration removes 99.999% of viruses
  • Optional antimicrobial wall guards inhibit bacterial growth
  • 7-year warranty
Catalog No. 50-283-8212 Supplier MSC Supplier No. 38866661
The XLERATOR hand dryer offers an 8-second dry time and custom image cover for personalized branding. Operating at 208-277V and 1490W, it provides energy and cost savings over paper towels, supporting environmental goals.
  • 8-second dry time
  • Adjustable speed, sound, and heat control
  • Visible service LED
  • 95% cost savings vs. paper towels
  • Reduces carbon footprint up to 88% vs. paper towels
  • Building Green Approved; contributes to WELL points & LEED credits
  • Optional electrostatic HEPA filtration removes 99.999% of viruses
  • Optional antimicrobial wall guards inhibit bacterial growth
  • 7-year warranty