Learn More
MSC Excel Dryer Hand Dryer, Automatic Sensor Operated, Surface Mounted, Adjustable Speed and Sound Control
Catalog No. 502836092
Encompass
The XLERATOR Hand Dryer is an original, patented hand dryer designed for high-performance and sustainability. It offers an 8-second dry time and significant cost savings compared to paper towels, making it a hygienic and environmentally friendly choice for various facilities.
  • Eight second dry time
  • Adjustable speed and sound control
  • Adjustable heat control
  • Externally visible service LED
  • Up to 95% cost savings versus paper towels
  • Up to 80% less energy than conventional hand dryers
  • No more maintenance and upkeep of paper towel waste
  • Building Green approved and helps contribute to WELL points and several LEED credits
  • HEPA filtration system proven to remove 99.999% of virus from the airstream
  • Antimicrobial wall guards inhibit bacterial growth
Catalog No. 50-283-6092 Supplier MSC Supplier No. 38866489
The XLERATOR Hand Dryer is an original, patented hand dryer designed for high-performance and sustainability. It offers an 8-second dry time and significant cost savings compared to paper towels, making it a hygienic and environmentally friendly choice for various facilities.
  • Eight second dry time
  • Adjustable speed and sound control
  • Adjustable heat control
  • Externally visible service LED
  • Up to 95% cost savings versus paper towels
  • Up to 80% less energy than conventional hand dryers
  • No more maintenance and upkeep of paper towel waste
  • Building Green approved and helps contribute to WELL points and several LEED credits
  • HEPA filtration system proven to remove 99.999% of virus from the airstream
  • Antimicrobial wall guards inhibit bacterial growth