Learn More
MSC Excel Dryer Hand Dryer, Brushed Stainless Steel, Automatic Sensor, Surface Mounted
Catalog No. 502838055
Encompass
This hand dryer with a brushed stainless steel cover provides fast and efficient hand drying. Designed for high-traffic environments, it features automatic sensor activation and surface mounting for easy installation.
  • Automatic sensor operation for hygienic use.
  • Brushed stainless steel cover for durability and aesthetics.
  • Surface mounted design for straightforward installation.
  • Provides an 8-second dry time.
  • Energy efficient with 1.7 Wh per use.
  • Powerful 5/8 hp motor with up to 24,000 RPM.
  • Operates with an average sound level of 62-75 dB(a).
  • Available with optional HEPA filtration system.
  • Available with optional 1.1 inch noise reduction nozzle.
  • ADA-compliant when used with an optional recess kit.
  • Backed by a 7-year limited warranty.
Catalog No. 50-283-8055 Supplier MSC Supplier No. 38866380
This hand dryer with a brushed stainless steel cover provides fast and efficient hand drying. Designed for high-traffic environments, it features automatic sensor activation and surface mounting for easy installation.
  • Automatic sensor operation for hygienic use.
  • Brushed stainless steel cover for durability and aesthetics.
  • Surface mounted design for straightforward installation.
  • Provides an 8-second dry time.
  • Energy efficient with 1.7 Wh per use.
  • Powerful 5/8 hp motor with up to 24,000 RPM.
  • Operates with an average sound level of 62-75 dB(a).
  • Available with optional HEPA filtration system.
  • Available with optional 1.1 inch noise reduction nozzle.
  • ADA-compliant when used with an optional recess kit.
  • Backed by a 7-year limited warranty.