Learn More
MSC Excel Dryer Hand Dryer, Brushed Stainless Steel, 1490 Watt
Catalog No. 502837912
Encompass
The XLERATOR Hand Dryer is a high-performance, automatic sensor-operated unit featuring a durable brushed stainless steel cover. Designed for efficiency and hygiene, this hand dryer delivers a powerful 1490 Watt operation, ensuring an 8-second dry time for various facilities.
  • 8-second dry time
  • Adjustable speed and sound control
  • Adjustable heat control (high, medium, low, and off)
  • 80% less energy than conventional hand dryers
  • 95% cost savings versus paper towels
  • Reduces maintenance and waste from paper towels
  • Contributes to reduced carbon footprint
  • Hygienic hand drying solution
  • Electrostatic HEPA filtration removes 99.999% of virus from airstream
  • Antimicrobial wall guards inhibit bacterial growth
  • Automatic sensor operated
  • Surface mounted
  • 7-year limited warranty
Catalog No. 50-283-7912 Supplier MSC Supplier No. 38866364
The XLERATOR Hand Dryer is a high-performance, automatic sensor-operated unit featuring a durable brushed stainless steel cover. Designed for efficiency and hygiene, this hand dryer delivers a powerful 1490 Watt operation, ensuring an 8-second dry time for various facilities.
  • 8-second dry time
  • Adjustable speed and sound control
  • Adjustable heat control (high, medium, low, and off)
  • 80% less energy than conventional hand dryers
  • 95% cost savings versus paper towels
  • Reduces maintenance and waste from paper towels
  • Contributes to reduced carbon footprint
  • Hygienic hand drying solution
  • Electrostatic HEPA filtration removes 99.999% of virus from airstream
  • Antimicrobial wall guards inhibit bacterial growth
  • Automatic sensor operated
  • Surface mounted
  • 7-year limited warranty