Learn More
MSC Excel Dryer XChanger Combo Kit, Paper Dispenser Retrofit, ADA Compliant, Stainless Steel
Catalog No. 502839643
Encompass
The XChanger Combo Kit provides a complete solution for retrofitting existing paper towel dispensers and waste receptacles with an XLERATOR or XLERATOReco hand dryer. Designed for easy installation, it covers the footprint of the previous dispenser, minimizing wall repair and helping to modernize restroom facilities while adhering to ADA guidelines.
  • Allows complete retrofit of paper dispenser and waste receptacle with a hand dryer.
  • Easily installed over existing paper dispenser space.
  • Does not damage existing wall surface.
  • Includes ADA recess kit.
  • Modernizes and cleans the restroom.
  • Minimizes or eliminates wall repair after installation.
  • Fabricated from 18 gauge 18-8 type 304 stainless steel with a #4 satin finish.
  • Features all welded construction.
Catalog No. 50-283-9643 Supplier MSC Supplier No. 77671550
The XChanger Combo Kit provides a complete solution for retrofitting existing paper towel dispensers and waste receptacles with an XLERATOR or XLERATOReco hand dryer. Designed for easy installation, it covers the footprint of the previous dispenser, minimizing wall repair and helping to modernize restroom facilities while adhering to ADA guidelines.
  • Allows complete retrofit of paper dispenser and waste receptacle with a hand dryer.
  • Easily installed over existing paper dispenser space.
  • Does not damage existing wall surface.
  • Includes ADA recess kit.
  • Modernizes and cleans the restroom.
  • Minimizes or eliminates wall repair after installation.
  • Fabricated from 18 gauge 18-8 type 304 stainless steel with a #4 satin finish.
  • Features all welded construction.