Learn More
MSC Excel Dryer XLERATOReco Hand Dryer, White Epoxy, 208-277V, 500 Watt
Catalog No. 502836006
Encompass
This XLERATOReco Hand Dryer is a high-speed, no-heat, and energy-efficient hand drying solution. It provides significant cost and environmental benefits by reducing reliance on paper towels and offering fast, hygienic drying for commercial settings.
  • High-speed, no-heat drying with 500 watts.
  • Achieves 95% cost savings compared to paper towels.
  • Reduces carbon footprint by up to 94% against paper towels.
  • Dries hands in 10 seconds.
  • Can install multiple units on a single circuit.
  • Requires minimal maintenance.
  • Uses 80% less energy than conventional hand dryers.
  • Available with electrostatic HEPA filtration.
  • Features a 7-year limited warranty.
  • Surface mounted design.
  • Automatic sensor operation.
Catalog No. 50-283-6006 Supplier MSC Supplier No. 38866323
This XLERATOReco Hand Dryer is a high-speed, no-heat, and energy-efficient hand drying solution. It provides significant cost and environmental benefits by reducing reliance on paper towels and offering fast, hygienic drying for commercial settings.
  • High-speed, no-heat drying with 500 watts.
  • Achieves 95% cost savings compared to paper towels.
  • Reduces carbon footprint by up to 94% against paper towels.
  • Dries hands in 10 seconds.
  • Can install multiple units on a single circuit.
  • Requires minimal maintenance.
  • Uses 80% less energy than conventional hand dryers.
  • Available with electrostatic HEPA filtration.
  • Features a 7-year limited warranty.
  • Surface mounted design.
  • Automatic sensor operation.