Learn More
MSC Excel Dryer XLERATOReco Hand Dryer, Brushed Stainless Steel, 500 W, 10 Second Dry Time
Catalog No. 502839308
Encompass
The XLERATOReco Hand Dryer is a high-speed, no-heat hand dryer designed for efficiency and hygiene. This model features a brushed stainless steel cover and provides a quick 10-second dry time, consuming only 500 watts of power. It offers significant cost and energy savings compared to traditional paper towels or conventional hand dryers.
  • 10-second dry time
  • 500 watts of power
  • 95% cost savings versus paper towels
  • Up to 94% carbon footprint reduction
  • Electrostatic HEPA filtration system removes 99.999% of viruses from airstream
  • 7-year limited warranty
  • 80% less energy than conventional hand dryers
  • Contributes to WELL points and LEED credits; EPD certified
  • Hygienic hand drying solution
  • Made in USA certified
  • Multi-voltage options (110-120V or 208-277V)
Catalog No. 50-283-9308 Supplier MSC Supplier No. 38866141
The XLERATOReco Hand Dryer is a high-speed, no-heat hand dryer designed for efficiency and hygiene. This model features a brushed stainless steel cover and provides a quick 10-second dry time, consuming only 500 watts of power. It offers significant cost and energy savings compared to traditional paper towels or conventional hand dryers.
  • 10-second dry time
  • 500 watts of power
  • 95% cost savings versus paper towels
  • Up to 94% carbon footprint reduction
  • Electrostatic HEPA filtration system removes 99.999% of viruses from airstream
  • 7-year limited warranty
  • 80% less energy than conventional hand dryers
  • Contributes to WELL points and LEED credits; EPD certified
  • Hygienic hand drying solution
  • Made in USA certified
  • Multi-voltage options (110-120V or 208-277V)