Learn More
OTT Hydromet Sutron - Xlink 500 No Modem, NEMA-4 Box
Catalog No. NC1919675
Encompass_Preferred
SUTRON’s XLink product family offers a cost-effective Wi-Fi enabled datalogger with data transmission via IRIDIUM or Cellular. Ideal for use with multiple digital and analog sensors supporting basic or advanced hydrologic or meteorologic monitoring applications. The product comes in two models, the 100 and 500, tailored to fit different application and budget requirements. The XLink 100 and XLink 500 carry over all the great capabilities from its predecessor family (CDMALink, HSPALink, GPRSLink, IridiumLink).
Catalog No. NC1919675 Supplier OTT Hydromet Supplier No. XLINK5001E
SUTRON’s XLink product family offers a cost-effective Wi-Fi enabled datalogger with data transmission via IRIDIUM or Cellular. Ideal for use with multiple digital and analog sensors supporting basic or advanced hydrologic or meteorologic monitoring applications. The product comes in two models, the 100 and 500, tailored to fit different application and budget requirements. The XLink 100 and XLink 500 carry over all the great capabilities from its predecessor family (CDMALink, HSPALink, GPRSLink, IridiumLink).