Learn More
Grainger XLR (F) Mic Connector 3pin
Catalog No. 502752417
Encompass_Preferred
The Grainger Approved XLR-F Mic Connector is a 3-pin female XLR connector. It is designed for balanced audio signals and features nickel-plated contacts for reliable connectivity. This XLR connector has a current rating of 7A and is compatible with 14-22AWG wire. It can be panel mounted and has a maximum temperature rating of 105°C. The connector body is made of black thermoplastic and it conforms to IEC 61076-2-103 standards. This is a professional grade audio connector suitable for microphones, mixing consoles and other pro audio equipment.
Catalog No. 50-275-2417 Supplier Grainger Supplier No. 14C228
The Grainger Approved XLR-F Mic Connector is a 3-pin female XLR connector. It is designed for balanced audio signals and features nickel-plated contacts for reliable connectivity. This XLR connector has a current rating of 7A and is compatible with 14-22AWG wire. It can be panel mounted and has a maximum temperature rating of 105°C. The connector body is made of black thermoplastic and it conforms to IEC 61076-2-103 standards. This is a professional grade audio connector suitable for microphones, mixing consoles and other pro audio equipment.