Learn More
Electron Microscopy Sciences Kodak Black Ribbon/X-Life Media
SDP
Catalog No. 5026777
Encompass
Media product for outstanding image stability and near-photographic quality continuous tone thermal prints. Feature excellent color reproduction, including superior flesh tones and durability. The unique Kodak XtraLife Ribbon overcoats prints with a rugged transparent layer which prevents destructive dye transfer as well as it resists fingerprints and fading. Only laminates to paper, when printing transparency material the laminate patch is bypassed.
Catalog No. 50-267-77 Supplier Electron Microscopy Sciences Supplier No. 89053
Media product for outstanding image stability and near-photographic quality continuous tone thermal prints. Feature excellent color reproduction, including superior flesh tones and durability. The unique Kodak XtraLife Ribbon overcoats prints with a rugged transparent layer which prevents destructive dye transfer as well as it resists fingerprints and fading. Only laminates to paper, when printing transparency material the laminate patch is bypassed.