Learn More
MOLTOX Molecular Toxicology Inc XLT4 Agar, 20 mL/plate, 10 plates/sleeve, Store in Refrigerator
SDP
Catalog No. 50948850
Encompass_Preferred
XLT4 (Xylose lysine tergitol 4) is a selective medium used in conjunction with the Niaproof 4 supplement to isolate non-typhi Salmonella from laboratory samples. Each lot of media from MOLTOX is tested for sterility, where applicable, and its ability to support the growth of the appropriate microorganism. All media are accompanied by GLP-compliant formulation and Quality Control certificate.
Catalog No. 50-948-850 Supplier MOLTOX Molecular Toxicology Inc Supplier No. 5140S122.140
XLT4 (Xylose lysine tergitol 4) is a selective medium used in conjunction with the Niaproof 4 supplement to isolate non-typhi Salmonella from laboratory samples. Each lot of media from MOLTOX is tested for sterility, where applicable, and its ability to support the growth of the appropriate microorganism. All media are accompanied by GLP-compliant formulation and Quality Control certificate.