Learn More
Biocare Medical, LLC XM Factor - 1.0 ml
SDP
Catalog No. 5083330
Encompass_Preferred
The XM Factor is a potent blocker for eliminating endogenous mouse IgG in mouse tissues. It has been specially designed to work with Biocare Rabbit-on-Rodent and Goat Polymer detection systems. XM Factor cannot be used with Mouse-on-Mouse AP or HRP polymer. By adding XM Factor to the polymer detection, endogenous mouse IgG is dramatically reduced and/or is completely eliminated. Biocare has also developed Rodent Block M that blocks for nonspecific background staining, and at the same time, also helps reduce endogenous mouse IgG. The combination of Rodent Block M and the XM Factor provide clean staining for mouse tissue protocols. It can also be used on an automated staining system.
Catalog No. 50-833-30 Supplier Biocare Medical, LLC Supplier No. XMF963C
The XM Factor is a potent blocker for eliminating endogenous mouse IgG in mouse tissues. It has been specially designed to work with Biocare Rabbit-on-Rodent and Goat Polymer detection systems. XM Factor cannot be used with Mouse-on-Mouse AP or HRP polymer. By adding XM Factor to the polymer detection, endogenous mouse IgG is dramatically reduced and/or is completely eliminated. Biocare has also developed Rodent Block M that blocks for nonspecific background staining, and at the same time, also helps reduce endogenous mouse IgG. The combination of Rodent Block M and the XM Factor provide clean staining for mouse tissue protocols. It can also be used on an automated staining system.