Learn More
New England Biolabs, Inc. XmaI – 2500 units
SDP
Catalog No. 50812096
Encompass
A restriction endonuclease that recognizes the sequence C^CCGG_G.
Catalog No. 50-812-096 Supplier New England Biolabs, Inc. Supplier No. R0180L
A restriction endonuclease that recognizes the sequence C^CCGG_G.