Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ xMAP™ Reporter Control Microspheres (CON2)

Catalog No. L100CON2
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
L100CON2 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. L100CON2 Supplier Invitrogen™ Supplier No. L100CON2
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

xMAP™ Reporter Control Microspheres (CON2), are polystyrene microspheres that are internally labeled with fluorescent dyes.

xMAP™ Reporter Control Microspheres (CON2), are polystyrene microspheres that are internally labeled with fluorescent dyes. These Control Microspheres are intended to verify the calibration and optical integrity for the Luminex 200™ System. Reporter Control Microspheres verify the reporter channel.

The Luminex™ 200™ System is sold separately.

TRUSTED_SUSTAINABILITY

Specifications

Channel Classification Reporter Channel (RP1)
For Use With (Equipment) Luminex™ Instruments, Luminex™ 200™ System
Content And Storage Store at 2°C to 8°C.
Product Line Luminex
Product Type Calibration Microspheres
Quantity 5 mL
Shipping Condition Wet Ice

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.