Learn More
Sigma Aldrich Fine Chemicals Biosciences Xmap Sheath Concentrate PL
Catalog No. 502900590
Encompass
The fLEXMAP 3D System(R) Luminex(R) 100 and Luminex 200(TM) Instruments require sheath fluid PLUS to serve as the delivery medium to transport the sample to the instruments optics.
Catalog No. 50-290-0590 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. 4050023
The fLEXMAP 3D System(R) Luminex(R) 100 and Luminex 200(TM) Instruments require sheath fluid PLUS to serve as the delivery medium to transport the sample to the instruments optics.