Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC XMD-17-51 TFA 10mM 1mL | 2436579-93-8 | 1 ML
SDP

Catalog No. 5000227585
Encompass_Preferred

XMD-17-51 (TFA) is a pyrimido-diazepinone small molecule that modulates protein kinases. It is supplied as the trifluoroacetate (TFA) salt and is provided as either a dry solid or a ready-to-use solution for biochemical and cell signaling research.

  • Modulates protein kinases for biochemical and cellular studies
  • Supplied as trifluoroacetate (TFA) salt, available as dry solid or solution
  • Typical solution: 10 mM in DMSO, 1 mL
  • High purity (LCMS 99.46%)
  • White to off-white solid appearance
  • Storage: 4°C for solid; in solvent -80°C (up to 6 months) or -20°C (up to 1 month)

Catalog No. 50-002-27585 Supplier Medchemexpress LLC Supplier No. HY117291A10MM 1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

XMD-17-51 (TFA) is a pyrimido-diazepinone small molecule that modulates protein kinases. It is supplied as the trifluoroacetate (TFA) salt and is provided as either a dry solid or a ready-to-use solution for biochemical and cell signaling research.

  • Modulates protein kinases for biochemical and cellular studies
  • Supplied as trifluoroacetate (TFA) salt, available as dry solid or solution
  • Typical solution: 10 mM in DMSO, 1 mL
  • High purity (LCMS 99.46%)
  • White to off-white solid appearance
  • Storage: 4°C for solid; in solvent -80°C (up to 6 months) or -20°C (up to 1 month)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.