Learn More
Medchemexpress LLC HY-113143A 50mg Medchemexpress, Galactose 1-phosphate Potassium salt CAS:19046-60-7 Purity:>98%
SDP
Catalog No. 501969231
Encompass_Preferred
Medchemexpress, HY-117291A 10mg XMD-17-51 Trifluoroacetate CAS: XMD-17-51 Trifluoroacetate is a pyrimido-diazepinone compound that is able to modulate protein kinases. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-9231 Supplier Medchemexpress LLC Supplier No. HY113143A50MG
Medchemexpress, HY-117291A 10mg XMD-17-51 Trifluoroacetate CAS: XMD-17-51 Trifluoroacetate is a pyrimido-diazepinone compound that is able to modulate protein kinases. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.