Learn More
Medchemexpress LLC HY-101243 5mg Medchemexpress, XMD16-5 CAS:1345098-78-3 Purity:>98%
SDP
Catalog No. 501932073
Encompass_Preferred
Medchemexpress, HY-101243 5mg XMD16-5 CAS:1345098-78-3 XMD16-5 is a potent TNK2 inhibitor with IC 50 values of 16 and 77 nM for the D163E and R806Q mutations, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-2073 Supplier Medchemexpress LLC Supplier No. HY1012435MG
Medchemexpress, HY-101243 5mg XMD16-5 CAS:1345098-78-3 XMD16-5 is a potent TNK2 inhibitor with IC 50 values of 16 and 77 nM for the D163E and R806Q mutations, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.