Learn More
Selleck Chemical LLC XMD8-92 10mg 1234480-50-2
SDP
Supplier:  Selleck Chemical LLC S752510MG
Encompass_Preferred
XMD8-92 is a potent and selective dual inhibitor of big map kinase (BMK1, ERK5) and bromodomain-containing proteins (BRDs, BET) with Kd of 80 nM and 170 nM for ERK5 and BRD4(1), respectively. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-136-4900