Learn More
Medchemexpress LLC XMD8-92 | 1234480-50-2 | 99.5% | C26H30N6O3 | 10 MM * 1 ML
SDP
Catalog No. 5000354517
Encompass_Preferred
XMD8-92 is a potent inhibitor of ERK5 (BMK1) and BRD4, with Kds of 80 nM and 190 nM, respectively. It also inhibits DCAMKL2, PLK4, and TNK1. This compound exhibits anti-cancer activity and is intended for research use only.
  • Potent ERK5 (BMK1) and BRD4 inhibitor
  • Kds of 80 nM for BMK1 and 190 nM for BRD4
  • Inhibits DCAMKL2, PLK4, and TNK1
  • Exhibits anti-cancer activity
  • For research use only
  • Inhibits proliferation of HMEC and cancer cells
  • Effectively inhibits BMK1 activation
  • Induces PML's downstream effector, p21
  • Significantly inhibits bFGF induced angiogenesis
Catalog No. 50-003-54517 Supplier Medchemexpress LLC Supplier No. HY1444310MM/1ML
XMD8-92 is a potent inhibitor of ERK5 (BMK1) and BRD4, with Kds of 80 nM and 190 nM, respectively. It also inhibits DCAMKL2, PLK4, and TNK1. This compound exhibits anti-cancer activity and is intended for research use only.
  • Potent ERK5 (BMK1) and BRD4 inhibitor
  • Kds of 80 nM for BMK1 and 190 nM for BRD4
  • Inhibits DCAMKL2, PLK4, and TNK1
  • Exhibits anti-cancer activity
  • For research use only
  • Inhibits proliferation of HMEC and cancer cells
  • Effectively inhibits BMK1 activation
  • Induces PML's downstream effector, p21
  • Significantly inhibits bFGF induced angiogenesis