Learn More
Selleck Chemical LLC XMD8-92
SDP
Catalog No. 5000004175
Encompass_Preferred
XMD8-92 is a potent and selective dual inhibitor of big map kinase (BMK1 ERK5) and bromodomain-containing proteins (BRDs BET) with Kd of 80 nM and 170 nM for ERK5 and BRD4(1) respectively
Catalog No. 50-000-04175 Supplier Selleck Chemical LLC Supplier No. S752550MG
XMD8-92 is a potent and selective dual inhibitor of big map kinase (BMK1 ERK5) and bromodomain-containing proteins (BRDs BET) with Kd of 80 nM and 170 nM for ERK5 and BRD4(1) respectively