Learn More
New England Biolabs, Inc. XmnI – 1000 units
SDP
Catalog No. 50812123
Encompass
A restriction endonuclease that recognizes the sequence GAANN_^NNTTC.
Catalog No. 50-812-123 Supplier New England Biolabs, Inc. Supplier No. R0194S
A restriction endonuclease that recognizes the sequence GAANN_^NNTTC.